Assay Details
Gene Symbol: LOC102123272 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_005555077.2 | XP_005555134.1 | 3 - 4 | 971 | 99 | |
GenBank mRNA | AB262776.2 | - | 3 - 4 | 316 | 99 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() |
Panther Classification:
