Assay Details
Gene Symbol: LOC102135463 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_005558575.2 | XP_005558632.1 | 4 - 5 | 1146 | 77 | |
XM_015452260.1 | XP_015307746.1 | 9 - 10 | 1167 | 77 | ||
XM_015452261.1 | XP_015307747.1 | 9 - 10 | 1147 | 77 | ||
XM_015452262.1 | XP_015307748.1 | 9 - 10 | 1202 | 77 | ||
XM_015452263.1 | XP_015307749.1 | 9 - 10 | 1358 | 77 | ||
XM_015452264.1 | XP_015307750.1 | 9 - 10 | 2190 | 77 | ||
XM_015452265.1 | XP_015307751.1 | 9 - 10 | 1140 | 77 | ||
XM_015452266.1 | XP_015307752.1 | 9 - 10 | 1182 | 77 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
