Assay Details
Gene Symbol: BAALC | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_005563849.2 | XP_005563906.1 | 2 - 3 | 1301 | 73 | |
GenBank mRNA | AB220478.1 | - | 2 - 3 | 260 | 73 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
