Assay Details
Gene Symbol: LOC102133480 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_005564188.2 | XP_005564245.1 | 2 - 3 | 726 | 103 | |
XM_015455286.1 | XP_015310772.1 | 3 - 4 | 377 | 103 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
