Assay Details
| Gene Symbol: NEFH | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015457323.1 | XP_015312809.1 | 1 - 2 | 948 | 65 | |
| GenBank mRNA | AB173983.1 | - | 1 - 2 | 925 | 65 | |
More Information
Set Membership: |
|

NEFH
Genomic Map