Assay Details
| Gene Symbol: FOXR1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_005579866.2 | XP_005579923.1 | 4 - 5 | 1196 | 92 | |
| XM_015435820.1 | XP_015291306.1 | 4 - 5 | 707 | 92 | ||
More Information
Set Membership: |
|

FOXR1
Genomic Map