Assay Details
| Gene Symbol: C15H9orf92 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_005581678.1 | XP_005581735.1 | 2 - 3 | 187 | 88 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

C15H9orf92| Gene Symbol: C15H9orf92 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_005581678.1 | XP_005581735.1 | 2 - 3 | 187 | 88 | |
Set Membership: |
|