Assay Details
| Gene Symbol: NOCT | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_005555932.2 | XP_005555989.1 | 2 - 3 | 1126 | 89 | |
| XM_005555934.2 | XP_005555991.1 | 2 - 3 | 481 | 89 | ||
More Information
Set Membership: |
|

NOCT
Genomic Map