Assay Details
Gene Symbol: CSRP3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_005578415.1 | XP_005578472.1 | 4 - 5 | 554 | 64 | |
XM_005578416.2 | XP_005578473.1 | 4 - 5 | 536 | 64 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
Panther Classification:
