Assay Details
Gene Symbol: CDKN2D | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_005587956.2 | XP_005588013.1 | 2 - 3 | 261 | 85 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |

Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: CDKN2D | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_005587956.2 | XP_005588013.1 | 2 - 3 | 261 | 85 |
Set Membership: |
![]() ![]() ![]() ![]() |