Assay Details
| Gene Symbol: HTR6 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_005544575.2 | XP_005544632.1 | 1 - 2 | 1344 | 77 | |
| GenBank mRNA | AY650371.1 | - | 1 - 2 | 63 | 77 | |
More Information
Set Membership: |
|

HTR6
Genomic Map