Assay Details
| Gene Symbol: LOC102123502 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015447058.1 | XP_015302544.1 | 10 - 11 | 2547 | 94 | |
| XM_015447059.1 | XP_015302545.1 | 10 - 11 | 1432 | 94 | ||
| XM_015447060.1 | XP_015302546.1 | 10 - 11 | 1353 | 94 | ||
| XM_015447061.1 | XP_015302547.1 | 10 - 11 | 1666 | 94 | ||
| XM_015447062.1 | XP_015302548.1 | 9 - 10 | 2373 | 94 | ||
| XM_015447063.1 | XP_015302549.1 | 8 - 9 | 2304 | 94 | ||
More Information
Set Membership: |
|

LOC102123502
Genomic Map