Assay Details
Gene Symbol: LOC102123198 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001319474.1 | NP_001306403.1 | 11 - 12 | 1066 | 81 | |
XM_015454704.1 | XP_015310190.1 | 11 - 12 | 1100 | 81 | ||
GenBank mRNA | X76637.1 | - | 1066 | 81 |

More Information
Set Membership: |
![]() ![]() |
Panther Classification:
