Assay Details
Gene Symbol: LOC102123198 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001319474.1 | NP_001306403.1 | 13 - 14 | 1350 | 120 | |
XM_015454704.1 | XP_015310190.1 | 13 - 14 | 1384 | 120 | ||
GenBank mRNA | X76637.1 | - | 1350 | 120 |

More Information
Set Membership: |
![]() ![]() |
Panther Classification:
