Assay Details
Gene Symbol: LOC102143735 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_015455822.1 | XP_015311308.1 | 11 - 12 | 6028 | 78 | |
XM_015455823.1 | XP_015311309.1 | 12 - 13 | 5196 | 78 | ||
XM_015455824.1 | XP_015311310.1 | 12 - 13 | 3073 | 78 | ||
XM_015455825.1 | XP_015311311.1 | 11 - 12 | 6028 | 78 | ||
XM_015455826.1 | XP_015311312.1 | 11 - 12 | 6028 | 78 | ||
XM_015455828.1 | XP_015311314.1 | 11 - 12 | 6028 | 78 | ||
XM_015455829.1 | XP_015311315.1 | 11 - 12 | 6028 | 78 | ||
XM_015455830.1 | XP_015311316.1 | 11 - 12 | 6025 | 78 | ||
XM_015455831.1 | XP_015311317.1 | 11 - 12 | 2526 | 78 | ||
XM_015455832.1 | XP_015311318.1 | 10 - 11 | 2220 | 78 | ||
XM_015455833.1 | XP_015311319.1 | 10 - 11 | 1502 | 78 |

More Information
Set Membership: |
![]() ![]() ![]() |
