Assay Details
| Gene Symbol: LOC102143735 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015455822.1 | XP_015311308.1 | 19 - 20 | 6533 | 127 | |
| XM_015455823.1 | XP_015311309.1 | 20 - 21 | 5701 | 127 | ||
| XM_015455824.1 | XP_015311310.1 | 20 - 21 | 3578 | 127 | ||
| XM_015455825.1 | XP_015311311.1 | 19 - 20 | 6533 | 127 | ||
| XM_015455826.1 | XP_015311312.1 | 19 - 20 | 6533 | 127 | ||
| XM_015455828.1 | XP_015311314.1 | 19 - 20 | 6533 | 127 | ||
| XM_015455829.1 | XP_015311315.1 | 19 - 20 | 6533 | 127 | ||
| XM_015455830.1 | XP_015311316.1 | 19 - 20 | 6530 | 127 | ||
| XM_015455831.1 | XP_015311317.1 | 19 - 20 | 3031 | 127 | ||
| XM_015455832.1 | XP_015311318.1 | 18 - 19 | 2725 | 127 | ||
| XM_015455833.1 | XP_015311319.1 | 18 - 19 | 2007 | 127 | ||
More Information
Set Membership: |
|

LOC102143735
Genomic Map