Assay Details
Gene Symbol: LOC102143735 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_015455822.1 | XP_015311308.1 | 20 - 21 | 6592 | 123 | |
XM_015455823.1 | XP_015311309.1 | 21 - 22 | 5760 | 123 | ||
XM_015455824.1 | XP_015311310.1 | 21 - 22 | 3637 | 123 | ||
XM_015455825.1 | XP_015311311.1 | 20 - 21 | 6592 | 123 | ||
XM_015455826.1 | XP_015311312.1 | 20 - 21 | 6592 | 123 | ||
XM_015455828.1 | XP_015311314.1 | 20 - 21 | 6592 | 123 | ||
XM_015455829.1 | XP_015311315.1 | 20 - 21 | 6592 | 123 | ||
XM_015455830.1 | XP_015311316.1 | 20 - 21 | 6589 | 123 | ||
XM_015455831.1 | XP_015311317.1 | 20 - 21 | 3090 | 123 | ||
XM_015455832.1 | XP_015311318.1 | 19 - 20 | 2784 | 123 | ||
XM_015455833.1 | XP_015311319.1 | 19 - 20 | 2066 | 123 |

More Information
Set Membership: |
![]() ![]() |
