Assay Details
Gene Symbol: LOC102143735 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_015455822.1 | XP_015311308.1 | 2 - 3 | 4617 | 81 | |
XM_015455823.1 | XP_015311309.1 | 3 - 4 | 3785 | 81 | ||
XM_015455824.1 | XP_015311310.1 | 3 - 4 | 1662 | 81 | ||
XM_015455825.1 | XP_015311311.1 | 2 - 3 | 4617 | 81 | ||
XM_015455826.1 | XP_015311312.1 | 2 - 3 | 4617 | 81 | ||
XM_015455828.1 | XP_015311314.1 | 2 - 3 | 4617 | 81 | ||
XM_015455829.1 | XP_015311315.1 | 2 - 3 | 4617 | 81 | ||
XM_015455830.1 | XP_015311316.1 | 2 - 3 | 4617 | 81 |
