Assay Details
| Gene Symbol: LOC102143735 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015455822.1 | XP_015311308.1 | 8 - 9 | 5851 | 91 | |
| XM_015455823.1 | XP_015311309.1 | 9 - 10 | 5019 | 91 | ||
| XM_015455824.1 | XP_015311310.1 | 9 - 10 | 2896 | 91 | ||
| XM_015455825.1 | XP_015311311.1 | 8 - 9 | 5851 | 91 | ||
| XM_015455826.1 | XP_015311312.1 | 8 - 9 | 5851 | 91 | ||
| XM_015455828.1 | XP_015311314.1 | 8 - 9 | 5851 | 91 | ||
| XM_015455829.1 | XP_015311315.1 | 8 - 9 | 5851 | 91 | ||
| XM_015455830.1 | XP_015311316.1 | 8 - 9 | 5848 | 91 | ||
| XM_015455831.1 | XP_015311317.1 | 8 - 9 | 2349 | 91 | ||
| XM_015455832.1 | XP_015311318.1 | 7 - 8 | 2043 | 91 | ||
| XM_015455833.1 | XP_015311319.1 | 7 - 8 | 1325 | 91 | ||

LOC102143735
Genomic Map