Assay Details
| Gene Symbol: FXYD4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_005565045.2 | XP_005565102.1 | 5 - 6 | 308 | 103 | |
| XM_005565046.2 | XP_005565103.1 | 5 - 6 | 335 | 103 | ||
| XM_005565047.2 | XP_005565104.1 | 5 - 6 | 308 | 103 | ||
| XM_005565048.2 | XP_005565105.1 | 5 - 6 | 335 | 103 | ||
| XM_005565050.2 | XP_005565107.1 | 5 - 6 | 308 | 103 | ||
| XM_015455961.1 | XP_015311447.1 | 5 - 6 | 232 | 103 | ||
| XM_015455963.1 | XP_015311449.1 | 5 - 6 | 227 | 103 | ||
More Information
Set Membership: |
|

FXYD4
Genomic Map