Assay Details
| Gene Symbol: EIF5A | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_005582727.1 | XP_005582784.1 | 3 - 4 | 402 | 77 | |
| XM_005582728.2 | XP_005582785.1 | 3 - 4 | 658 | 77 | ||
| XM_005582729.2 | XP_005582786.1 | 3 - 4 | 494 | 77 | ||
| XM_015437571.1 | XP_015293057.1 | 3 - 4 | 307 | 77 | ||
| XM_015437572.1 | XP_015293058.1 | 3 - 4 | 307 | 77 | ||
More Information
Set Membership: |
|

EIF5A
Genomic Map