Assay Details
Gene Symbol: CDRT4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_005582945.2 | XP_005583002.1 | 3 - 4 | 308 | 123 | |
XM_005582946.2 | XP_005583003.1 | 5 - 6 | 687 | 123 | ||
XM_015437672.1 | XP_015293158.1 | 3 - 4 | 284 | 123 | ||
XM_015437673.1 | XP_015293159.1 | 3 - 4 | 308 | 123 | ||
XM_015437674.1 | XP_015293160.1 | 3 - 4 | 256 | 123 |

More Information
Additional Information:
This assay is in the 5'UTR region of the XM_005582946.2, XM_015437673.1, XM_015437674.1 transcripts.
Set Membership: |
![]() ![]() ![]() ![]() |
