Assay Details
| Gene Symbol: ZNF571 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_005589037.2 | XP_005589094.1 | 4 - 5 | 907 | 72 | |
| XM_005589038.2 | XP_005589095.1 | 4 - 5 | 775 | 72 | ||
| XM_015441217.1 | XP_015296703.1 | 4 - 5 | 347 | 72 | ||
More Information
Set Membership: |
|

ZNF571
Genomic Map