Assay Details
Gene Symbol: C1GALT1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_005550140.2 | XP_005550197.1 | 2 - 3 | 525 | 114 | |
XM_015447709.1 | XP_015303195.1 | 1 - 2 | 236 | 114 | ||
XM_015447710.1 | XP_015303196.1 | 3 - 4 | 625 | 114 | ||
XM_015447711.1 | XP_015303197.1 | 2 - 3 | 423 | 114 | ||
XM_015447712.1 | XP_015303198.1 | 1 - 2 | 236 | 114 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Panther Classification:
