Assay Details
| Gene Symbol: GPX8 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_005556896.2 | XP_005556953.1 | 2 - 3 | 531 | 93 | |
| XM_015451511.1 | XP_015306997.1 | 2 - 3 | 378 | 93 | ||
More Information
Set Membership: |
|

GPX8
Genomic Map