Assay Details
Gene Symbol: SFTA3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_015453720.1 | XP_015309206.1 | 3 - 4 | 3195 | 105 |

More Information
Set Membership: |
![]() ![]() ![]() |

Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: SFTA3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_015453720.1 | XP_015309206.1 | 3 - 4 | 3195 | 105 |
Set Membership: |
![]() ![]() ![]() |