Assay Details
| Gene Symbol: C7H14orf180 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_005562337.2 | XP_005562394.1 | 5 - 6 | 863 | 66 | |
| XM_005562339.2 | XP_005562396.1 | 5 - 6 | 1893 | 66 | ||
| XM_005562340.2 | XP_005562397.1 | 5 - 6 | 1871 | 66 | ||
| XM_005562341.2 | XP_005562398.1 | 3 - 4 | 736 | 66 | ||
| XM_015454252.1 | XP_015309738.1 | 5 - 6 | 805 | 66 | ||
| XM_015454253.1 | XP_015309739.1 | 3 - 4 | 1772 | 66 | ||
More Information
Set Membership: |
|

C7H14orf180
Genomic Map