Assay Details
Gene Symbol: LOC102132378 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_005570065.2 | XP_005570122.1 | 7 - 8 | 2596 | 102 | |
XM_015430345.1 | XP_015285831.1 | 7 - 8 | 2596 | 102 |

More Information
Set Membership: |
![]() ![]() |
Panther Classification:
