Assay Details
| Gene Symbol: PRRG4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_005578266.2 | XP_005578323.1 | 5 - 6 | 852 | 104 | |
| XM_005578267.2 | XP_005578324.2 | 5 - 6 | 691 | 104 | ||
More Information
Set Membership: |
|

PRRG4
Genomic Map