Assay Details
Gene Symbol: LOC102136393 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_015437764.1 | XP_015293250.1 | 10 - 11 | 2052 | 90 | |
XM_015437765.1 | XP_015293251.1 | 9 - 10 | 887 | 90 | ||
XM_015437766.1 | XP_015293252.1 | 10 - 11 | 1001 | 90 |

More Information
Set Membership: |
![]() ![]() |
Panther Classification:
