Assay Details
Gene Symbol: LOC102136393 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001319494.1 | NP_001306423.1 | 14 - 15 | 1305 | 91 | |
XM_015437764.1 | XP_015293250.1 | 14 - 15 | 2440 | 91 | ||
XM_015437765.1 | XP_015293251.1 | 13 - 14 | 1275 | 91 | ||
GenBank mRNA | KP731383.1 | - | 14 - 15 | 1305 | 91 |

More Information
Set Membership: |
![]() ![]() |
Panther Classification:
