Assay Details
Gene Symbol: LOC102136393 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001319494.1 | NP_001306423.1 | 6 - 7 | 544 | 93 | |
XM_015437764.1 | XP_015293250.1 | 6 - 7 | 1679 | 93 | ||
XM_015437765.1 | XP_015293251.1 | 6 - 7 | 628 | 93 | ||
XM_015437766.1 | XP_015293252.1 | 6 - 7 | 628 | 93 | ||
GenBank mRNA | KP731383.1 | - | 6 - 7 | 544 | 93 |

More Information
Set Membership: |
![]() ![]() |
Panther Classification:
