Assay Details
| Gene Symbol: CLEC10A | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015437550.1 | XP_015293036.1 | 3 - 4 | 1381 | 63 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

CLEC10A| Gene Symbol: CLEC10A | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015437550.1 | XP_015293036.1 | 3 - 4 | 1381 | 63 | |
Set Membership: |
|