Assay Details
Gene Symbol: WFIKKN2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_005583730.2 | XP_005583787.1 | 1 - 2 | 1091 | 98 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |

Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: WFIKKN2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_005583730.2 | XP_005583787.1 | 1 - 2 | 1091 | 98 |
Set Membership: |
![]() ![]() ![]() ![]() |