Assay Details
Gene Symbol: ITPRIPL2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_005591354.2 | XP_005591411.1 | 1 | 1824 | 109 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() |

Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: ITPRIPL2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_005591354.2 | XP_005591411.1 | 1 | 1824 | 109 |
Set Membership: |
![]() ![]() ![]() ![]() ![]() |