Assay Details
| Gene Symbol: LOC102137491 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_005594526.2 | XP_005594583.1 | 1 - 2 | 1438 | 84 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

LOC102137491| Gene Symbol: LOC102137491 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_005594526.2 | XP_005594583.1 | 1 - 2 | 1438 | 84 | |
Set Membership: |
|