Assay Details
| Gene Symbol: ZNF480 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015441991.1 | XP_015297477.1 | 3 - 4 | 331 | 69 | |
| XM_015441992.1 | XP_015297478.1 | 3 - 4 | 774 | 69 | ||
| XM_015441993.1 | XP_015297479.1 | 3 - 4 | 270 | 69 | ||
| XM_015441994.1 | XP_015297480.1 | 3 - 4 | 271 | 69 | ||
| XM_015441995.1 | XP_015297481.1 | 2 - 3 | 170 | 69 | ||
More Information
Set Membership: |
|

ZNF480
Genomic Map