Assay Details
Gene Symbol: RAX2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_005587531.1 | XP_005587588.1 | 2 - 3 | 277 | 111 | |
XM_015440231.1 | XP_015295717.1 | 2 - 3 | 534 | 111 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
Panther Classification:
