Assay Details
| Gene Symbol: LOC102119195 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_005549451.2 | XP_005549508.2 | 3 - 4 | 400 | 76 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

LOC102119195| Gene Symbol: LOC102119195 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_005549451.2 | XP_005549508.2 | 3 - 4 | 400 | 76 | |
Set Membership: |
|