Assay Details
| Gene Symbol: TPT1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_005585783.2 | XP_005585840.1 | 6 | 1217 | 116 | |
| GenBank mRNA | AB170187.1 | - | 6 | 1017 | 116 | |
More Information
Set Membership: |
|

TPT1
Genomic Map