Assay Details
| Gene Symbol: CMPK1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_005543448.1 | XP_005543505.1 | 6 | 1526 | 120 | |
| XM_015436317.1 | XP_015291803.1 | 6 | 1483 | 120 | ||
| GenBank mRNA | AB220489.1 | - | 2 | 852 | 120 | |
More Information
Set Membership: |
|

CMPK1
Genomic Map