Assay Details
| Gene Symbol: KHDC1L | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_005552562.2 | XP_005552619.1 | 3 | 1186 | 80 | |
| Gene Symbol: LOC102145562 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015454527.1 | XP_015310013.1 | 1 | 697 | 80 | |
More Information
Set Membership: |
|

KHDC1L
Genomic Map