Assay Details
Gene Symbol: MPHOSPH6 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_005592646.2 | XP_005592703.1 | 5 | 717 | 146 | |
XM_005592647.2 | XP_005592704.1 | 6 | 746 | 146 | ||
XM_005592648.2 | XP_005592705.1 | 5 | 732 | 146 | ||
XM_015443217.1 | XP_015298703.1 | 5 | 1181 | 146 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
