Assay Details
Gene Symbol: CDRT15L2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_005583082.2 | XP_005583139.1 | 2 | 1359 | 117 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() |

Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: CDRT15L2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_005583082.2 | XP_005583139.1 | 2 | 1359 | 117 |
Set Membership: |
![]() ![]() ![]() ![]() ![]() |