Assay Details
| Gene Symbol: C4H6orf163 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001284172.1 | NP_001271101.1 | 3 - 4 | 592 | 146 | |
| XM_015448836.1 | XP_015304322.1 | 4 - 5 | 2578 | 146 | ||
| GenBank mRNA | AB070040.1 | - | 3 - 4 | 592 | 146 | |
More Information
Set Membership: |
|

C4H6orf163
Genomic Map