Assay Details
Gene Symbol: C4H6orf163 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001284172.1 | NP_001271101.1 | 4 - 5 | 803 | 101 | |
XM_005552408.2 | XP_005552465.1 | 3 - 4 | 927 | 101 | ||
XM_015448836.1 | XP_015304322.1 | 5 - 6 | 2789 | 101 | ||
GenBank mRNA | AB070040.1 | - | 4 - 5 | 803 | 101 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
