Assay Details
| Gene Symbol: PHETA1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001285325.1 | NP_001272254.1 | 3 | 3522 | 100 | |
| XM_005572264.2 | XP_005572321.1 | 3 | 100 | |||
| GenBank mRNA | AB168244.1 | - | 3 | 3522 | 100 | |
More Information
Set Membership: |
|

PHETA1
Genomic Map