Assay Details
| Gene Symbol: CXCL1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001283494.1 | NP_001270423.1 | 3 - 4 | 324 | 73 | |
| GenBank mRNA | AB262775.1 | - | 3 - 4 | 324 | 73 | |
More Information
Set Membership: |
|

CXCL1
Genomic Map