Assay Details
Gene Symbol: BANF1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_005577199.2 | XP_005577256.1 | 2 - 3 | 264 | 105 | |
XM_005577200.2 | XP_005577257.1 | 2 - 3 | 216 | 105 | ||
XM_005577202.2 | XP_005577259.1 | 2 - 3 | 252 | 105 | ||
XM_005577203.2 | XP_005577260.1 | 2 - 3 | 619 | 105 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
