Assay Details
Gene Symbol: GSTO2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001319441.1 | NP_001306370.1 | 2 - 3 | 143 | 74 | |
XM_005566369.2 | XP_005566426.1 | 3 - 4 | 1757 | 74 | ||
XM_005566370.2 | XP_005566427.1 | 3 - 4 | 1762 | 74 | ||
GenBank mRNA | KC734503.1 | - | 2 - 3 | 143 | 74 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
